▸
Organic compounds
▸
Organonitrogen Compounds
▸
Hydrazines and derivatives
▸
Mineral oil, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Mineral oil, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0215169491
1 L
Catalog number
MPB0215169491
Price (SGD)
103.00
Each
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (SGD)
103.00
Each
Add to cart
Specifications
Chemical Name or Material
Mineral oil
CAS
8042-47-5
Color
Colorless
Density
0.81 to 0.894 at 15°C
Molecular Formula
C16H10N2Na2O7S2
View more