Leupeptin, MP Biomedicals™
Leupeptin, MP Biomedicals™
Actual product may vary
Leupeptin, MP Biomedicals™
Leupeptin, MP Biomedicals™
MP Biomedicals
Leupeptin, MP Biomedicals™
Inhibits trypsin-like proteases and some cysteine proteases. MP Biomedicals' Leupeptin is a modified tripeptide reversible protease inhibitor. Little to no inhibition is seen against pepsin, cathepsins A and D, and alpha-chymotrypsin.
Have Questions?
Catalog NumberQuantity
MPB02151553055mg
Catalog number MPB0215155305
Price (SGD)
194.00
EA
Add to cart
Quantity:
5mg
Price (SGD)
194.00
EA
Add to cart
When adjusted for molarity, all three salt forms are equally effective; however, the hydrochloride salt is usually less invasive in biological settings.
Inhibits trypsin-like proteases and some cysteine proteases including: endoproteinase Lys-C, kallikrein, papain, thrombin, cathepsin B, cathepsins H and L, trypsin, calpain, trypsin8 and plasmin.
Specifications
CAS103476-89-7
FormatWhite Powder
Molecular Weight (g/mol)427.57
Quantity5mg
Unit SizeEA
Contents & Storage
Store at −20°C, desiccate, protect from light.