1,1,1,3,3,3-Hexafluoroisopropanol, >99%, MP Biomedicals™
1,1,1,3,3,3-Hexafluoroisopropanol, >99%, MP Biomedicals™
1,1,1,3,3,3-Hexafluoroisopropanol, >99%, MP Biomedicals™
1,1,1,3,3,3-Hexafluoroisopropanol, >99%, MP Biomedicals™
MP Biomedicals
1,1,1,3,3,3-Hexafluoroisopropanol, >99%, MP Biomedicals™
1,1,1,3,3,3-Hexafluoro-Isopropanol is a speciality solvent effective for solubilizing a wide range of polymers, including those that are not soluble in the most common organic solvents.
Have Questions?
Catalog NumberQuantity
MPB0215124580100 mL
Catalog number MPB0215124580
Price (SGD)
452.00
EA
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (SGD)
452.00
EA
Add to cart
Specifications
Chemical Name or Material1,1,1,3,3,3-Hexafluoro-isopropanol
Name NoteFor GC analysis
CAS29463-77-2
ColorColorless
Recommended StorageStore at room temperature (15°C to 30°C).
View more
  • 1,1,1,3,3,3-Hexafluoro-Isopropanol is a speciality solvent effective for solubilizing a wide range of polymers, including those that are not soluble in the most common organic solvents, such as:
    • Polyamides, polyacrylonitriles, polyacetals, polyesters (e.g. polyglycolide), and polyketones.
  • It has also found use in biochemistry to solubilize peptides and to monomerize β-sheet protein aggregates.
  • It can be used as an acid in volatile buffers for ion pair HPLC - mass spectrometry of nucleic acids.
  • 1,1,1,3,3,3-Hexafluoro-2-propanol was used for preparing hexafluoroalcohol-functionalized methacrylate polymers for lithographic/nanopatterning materials.
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.