Check Order Status
Formic Acid, Free Acid, >97%, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Formic Acid, Free Acid, >97%, MP Biomedicals™
Sodium formate is used in several fabric dyeing and printing processes.
Have Questions?
Catalog Number
Quantity
MPB0215116290
500 mL
Catalog number
MPB0215116290
Price (SGD)
-
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or Material
Formic Acid, free acid
CAS
64-18-6
Melting Point
253°C
Color
Colorless
Assay Percent Range
>97%
View more
It is also used as a buffering agent for strong mineral acids to increase their pH, and as a food additive (E237).