Check Order Status
Diaphorase from
Clostridium kluyveri
MP Biomedicals
Diaphorase from
Clostridium kluyveri
Used to assess the protein-protein interactions in assembly of lipoic acid on the 2-oxoacid dehydrogenases of aerobic metabolism
Have Questions?
Catalog Number
Quantity
MPB0215084301
1.8 kU
Catalog number
MPB0215084301
Price (SGD)
1,268.00
EA
Add to cart
Quantity:
1.8 kU
Price (SGD)
1,268.00
EA
Add to cart
Specifications
Color
Dark Golden
Quantity
1.8 kU
Product Type
Diaphorase, Lipoamide Dehydrogenase, Lipoyl Dehydrogenase
Form
Powder
Unit Size
EA
Contents & Storage
0°C