Brilliant Green, MP Biomedicals™
Brilliant Green, MP Biomedicals™
Brilliant Green, MP Biomedicals™
Brilliant Green, MP Biomedicals™
MP Biomedicals
Brilliant Green, MP Biomedicals™
Also called emerald green or malachite green. Brilliant Green, MP Biomedicals is one of the triarylmethane dyes of the malachite-green series. It occurs as small, shiny, golden crystals soluble in water or alcohol.
Have Questions?
Catalog NumberQuantity
MPB021505162525 g
Catalog number MPB0215051625
Price (SGD)
60.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (SGD)
60.00
EA
Add to cart
Specifications
CAS633-03-4
Recommended StorageRoom Temperature (15–30°C).
Molecular FormulaC27H34N2O4S
SynonymAstradiamant green GX, Basic Green 1, Diamond Green, Emerald Green, Ethyl Green, Malachite Green G, Solid Green JO
Solubility InformationSoluble in water (1mg/mL), ethanol, methanol.
View more
Brilliant green is used in dilute solution as a topical antiseptic. It is effective against gram-positive microorganisms. It has also been used to dye silk and wool.