Check Order Status
Fast Blue B Salt , MP Biomedicals, LLC
Element Search
Structure Search
MP Biomedicals
Fast Blue B Salt , MP Biomedicals, LLC
Fast Blue B Salt is a stain that is used for the determination of various compounds.
Have Questions?
Catalog Number
Quantity
MPB0215020180
100 g
Catalog number
MPB0215020180
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Absorbance
λmax (water): 208 to 211, 305 to 308, 369 to 374 nm
Packaging
Glass
Color
Yellow
Quantity
100 g
Unit Size
EA
Fast blue B salt is useful for TLC of Aflatoxins
Fast Blue B is a diazonium salt used as a tissue stain
It has been used to demonstrate esterase activity
Histochemically it has stained enterochromaffin cells and cells that have gone under free radical oxidative damage