Check Order Status
Paraformaldehyde MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Paraformaldehyde MP Biomedicals
Have Questions?
Catalog Number
Quantity
MPB0215014601
1 kg
Catalog number
MPB0215014601
Price (SGD)
289.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (SGD)
289.00
EA
Add to cart
Specifications
Chemical Name or Material
Paraformaldehyde
CAS
30525-89-4
Density
1.4 g/mL
Molecular Formula
CH2O
Quantity
1 kg
View more