Check Order Status
Caffeine Anhydrous MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Caffeine Anhydrous MP Biomedicals
Have Questions?
Catalog Number
Quantity
MPB0215011480
100 g
Catalog number
MPB0215011480
Price (SGD)
78.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (SGD)
78.00
EA
Add to cart
Specifications
Chemical Name or Material
Caffeine Anhydrous
Name Note
Anhydrous
CAS
58-08-2
Color
White
Recommended Storage
Store at room temperature (15° to 30°C).
View more