Check Order Status
5-Fluorodeoxyuridine, ≥98%, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
5-Fluorodeoxyuridine, ≥98%, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0210555125
25 mg
Catalog number
MPB0210555125
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mg
Request bulk or custom format
Specifications
Chemical Name or Material
5-Fluorodeoxyuridine
CAS
50-91-9
Color
White
Assay Percent Range
≥98%
Molecular Formula
C9H11FN2O5
View more