Check Order Status
Actinomycin D Crystalline MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Actinomycin D Crystalline MP Biomedicals
Have Questions?
Catalog Number
Quantity
MPB0210465805
5 mg
Catalog number
MPB0210465805
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 mg
Request bulk or custom format
Specifications
Chemical Name or Material
Actinomycin D
Type
Actinomycin D
Physical Form
Crystalline Powder
CAS
50-76-0
Storage Note 1
Protect from light.
View more