▸
Cell Culture Media, Supplements, and Reagents
▸
Cell Culture Reagents, Antibiotics and Supplements
▸
Nutritional Supplements
▸
Yeast Extract Powder
MP Biomedicals
Yeast Extract Powder
A vacuum-dried extract concentrate of Baker's yeast containing the B-complex factors of approximately three times the weight of ordinary dry yeast.
Have Questions?
Change view
Catalog Number
Quantity
MPB0210330391
1 kg
MPB0210330390
500 g
2 Options
Catalog number
MPB0210330391
Price (SGD)
423.00
Each
Add to cart
Quantity:
1 kg
Price (SGD)
423.00
Each
Add to cart
Specifications
pH
5 to 5.5
Quantity
1 kg
Unit Size
Each
Contents & Storage
15° to 30°C