Vitamin B12, MP Biomedicals™
Vitamin B12, MP Biomedicals™
Vitamin B12, MP Biomedicals™
MP Biomedicals
Vitamin B12, MP Biomedicals™
Plays a role in myelin synthesis and is used an antipernicious anemia factor. Vitamin B12, MP Biomedicals (cobalamin) is a group of chemically-related cobalt-containing molecules involved in cell processes such as DNA synthesis, fatty acid synthesis, ener
Have Questions?
Catalog NumberQuantity
MPB02103271.2250 mg
Catalog number MPB02103271.2
Price (SGD)
110.00
Each
Quantity:
250 mg
Request bulk or custom format
Price (SGD)
110.00
Each
Specifications
CAS68-19-9
Melting Point>300°C (literature)
Recommended Storage4°C, desiccate.
Molecular FormulaC63H88CoN14O14P
MDL NumberMFCD00151092
View more
The physiologically active forms of vitamin B12 include methylcobalamin and adenosylcobalamin, whereas hydroxocobalamin (vitamin B12a, OHCbl) and cyanocobalamin (CNCbl) are storage and delivery forms.
Biosynthesis of the basic structure of the vitamin is accomplished only by bacteria (which usually produce hydroxocobalamin). Bacteria-derived hydroxocobalamin (OHCbl) and CNCbl are converted in humans to useful coenzyme forms to support metabolic processes, such as mitochondrial methylmalonylcoenzyme A mutase conversion of methylmalonic acid (MMA) to succinate to link lipid and carbohydrate metabolism, and activation of methionine synthase, the rate limiting step in the synthesis of methionine.
  • Vitamin B12 plays a role in the synthesis of myelin
  • It is a cofactor for two enzymes, methionine synthase and L-methylmalonyl-CoA mutase
  • The compound is also used as an antipernicious anemia factor