▸
Facility Safety and Maintenance
▸
Cleaning Supplies and Equipment
▸
Cleaners and Disinfectants
▸
Laboratory Surfactants and Wetting Agents
▸
Tween™80, ∽75% oleic acid, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Tween™80, ∽75% oleic acid, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0210317090
0.5 L
Catalog number
MPB0210317090
Price (SGD)
117.00
Each
Add to cart
Quantity:
0.5 L
Request bulk or custom format
Price (SGD)
117.00
Each
Add to cart
Specifications
Chemical Name or Material
Tween 80
Color
Yellow
CAS
9005-70-3
Physical Form
Liquid
Recommended Storage
Store at room temperature.
View more