Check Order Status
L-(-)-Sorbose 99+% MP Biomedicals
Element Search
Structure Search
MP Biomedicals
L-(-)-Sorbose 99+% MP Biomedicals
Have Questions?
Catalog Number
Quantity
MPB0210293980
100 g
Catalog number
MPB0210293980
Price (SGD)
435.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (SGD)
435.00
EA
Add to cart
Specifications
Chemical Name or Material
L-(-)-Sorbose
CAS
87-79-6
Assay Percent Range
99+%
Molecular Formula
C6H12O6
Quantity
100 g
View more