Check Order Status
Salicin, MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Salicin, MP Biomedicals
Salicin is a substrate for β-glucosidase
Have Questions?
Catalog Number
Quantity
MPB0210283380
100 g
Catalog number
MPB0210283380
Price (SGD)
939.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (SGD)
939.00
EA
Add to cart
Specifications
Chemical Name or Material
Salicin
CAS
138-52-3
EINECS Number
205-331-6
Molecular Formula
C13H18O7
MDL Number
MFCD00006590
View more