Check Order Status
D-(+)-Raffinose pentahydrate, 98%, ≥98%, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
D-(+)-Raffinose pentahydrate, 98%, ≥98%, MP Biomedicals™
Have Questions?
Change view
Quantity:
500 g
25 g
100 g
Catalog number
MPB0210279790
Price (SGD)
1,729.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (SGD)
1,729.00
EA
Add to cart
Specifications
Chemical Name or Material
D-(+)-Raffinose pentahydrate
Name Note
For HPLC analysis
CAS
17629-30-0
Color
White
Density
1.465g/mL
View more