Check Order Status
Phenolphthalein, U.S.P./N.F. grade, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Phenolphthalein, U.S.P./N.F. grade, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0210260883
250 g
Catalog number
MPB0210260883
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or Material
Phenolphthalein
CAS
77-09-8
Recommended Storage
Store at 2° to 8°C.
EINECS Number
201-004-7
Assay Percent Range
U.S.P./N.F. grade
View more