Pepsin
Pepsin
Actual product may vary
MP Biomedicals
Pepsin
Pepsin is a peptidase used to digest proteins and is commonly used in the preparation of Fab fragments from antibodies. Pepsin, from porcine gastric mucosa, has been used to hydrolyze dry cervical samples in mice
Have Questions?
Catalog NumberQuantity
MPB02102598911 kg
Catalog number MPB0210259891
Price (SGD)
3,904.00
EA
Add to cart
Quantity:
1 kg
Price (SGD)
3,904.00
EA
Add to cart
Pepsin is a peptidase and the major proteolytic enzyme produced in the stomach. It is used to digest proteins through the cleavage of interior peptide linkages. It is commonly used in the preparation of Fab fragments from antibodies. Pepsin from porcine gastric mucosa has been used to hydrolyze dry cervical samples in mice.
  • Pepsin hydrolyzes peptide bonds, not amide or ester linkages
  • Pepsin cleaves peptides with an aromatic acid on either side of the peptide bond
  • Sulfur-containing amino acids increase susceptibility to hydrolysis when they are close to the peptide bond
  • Pepsin preferentially cleaves at the carboxyl side of phenylalanine and leucine and at the carboxyl side of glutamic acid residues
  • Cleaves Phe-Val, Gln-His, Glu-Ala, Ala-Leu, Leu-Tyr, Tyr-Leu, Gly-Phe, Phe-Phe and Phe-Tyr bonds in the β chain of insulin
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.
Specifications
Product TypeProtease
CAS9001-75-6
For Use With (Application)Used as a proteolytic enzyme, Used to break down the proteins into their components, i.e. peptides and aminoacids
Quantity1 kg
Unit SizeEA