Methyl P-Hydroxybenzoate, MP Biomedicals™
Methyl P-Hydroxybenzoate, MP Biomedicals™
Methyl P-Hydroxybenzoate, MP Biomedicals™
Methyl P-Hydroxybenzoate, MP Biomedicals™
MP Biomedicals
Methyl P-Hydroxybenzoate, MP Biomedicals™
Used as a preservative in foods, beverages and cosmetics. Methyl P-Hydroxybenzoate, MP Biomedicals is the methyl ester of p-hydroxybenzoic acid. It can be used to inhibit yeast growth and can also be used as an antimicrobial agent for Gram-positive anaero
Have Questions?
Catalog NumberQuantity
MPB02102341.1100 g
Catalog number MPB02102341.1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
CAS99-76-3
Melting Point124°C to 131°C
Recommended StorageStore at Room Temperature (15–30°C)
Boiling Point270°C to 280°C
Molecular FormulaC8H8O3
View more
Methyl P-Hydroxybenzoate must be suspended in ethanol first, since it does not dissolve readily in water. It is indicated to have some pheromonal properties..