D-Mannitol, MP Biomedicals
D-Mannitol, MP Biomedicals
D-Mannitol, MP Biomedicals
D-Mannitol, MP Biomedicals
MP Biomedicals
D-Mannitol, MP Biomedicals
Have Questions?
Catalog NumberQuantity
MPB02102248.5500 g
Catalog number MPB02102248.5
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialD-MANNITOL
CAS69-65-8
Melting Point163°C to 172°C
Recommended StorageRoom Temperature
Boiling Point290°C to 295°C (3.5mm)(literature)
View more
  • D-Mannitol used with boric acid in the manufacturing of dry electrolytic condensers for radio applications
  • In pharmacy as excipient and diluent for solids and liquids
  • In analytical chemistry for boron determinations
  • In the manufuring of mannitol hexanitrate
  • It is used as an anticaking and free-flow agent, flavoring agent, lubricant and release agent, stabilizer and thickener and nutritive sweetener in the food industry
  • Also used as a diuretic