Check Order Status
D-(+)-Maltose monohydrate, 95 to 98%, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
D-(+)-Maltose monohydrate, 95 to 98%, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0210224180
100 g
Catalog number
MPB0210224180
Price (SGD)
133.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (SGD)
133.00
EA
Add to cart
Specifications
Chemical Name or Material
D-(+)-Maltose monohydrate
CAS
6363-53-7
Color
White
Recommended Storage
Store at room temperature.
Density
1.54g/mL
View more
Molecular biology