Check Order Status
Isopropyl-β-D-Thiogalactopyranoside, (Crystalline),MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Isopropyl-β-D-Thiogalactopyranoside, (Crystalline),MP Biomedicals
Dioxane-free β-Galactosidase inducer
Have Questions?
Catalog Number
Quantity
MPB0210210105
5 g
Catalog number
MPB0210210105
Price (SGD)
587.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (SGD)
587.00
EA
Add to cart
Specifications
Chemical Name or Material
Isopropyl-β-D-Thiogalactopyranoside
CAS
367-93-1
Recommended Storage
0°C
Molecular Formula
C9H18O5S
MDL Number
MFCD00063273
View more