Check Order Status
Indole-3-propionic acid, ∽99%, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Indole-3-propionic acid, ∽99%, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0210204801
1 g
Catalog number
MPB0210204801
Price (SGD)
76.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (SGD)
76.00
EA
Add to cart
Specifications
Chemical Name or Material
Indole-3-propionic acid
Name Note
Molecular biology
CAS
830-96-6
Color
Cream
Recommended Storage
Store at -0°C
View more