Check Order Status
Imidazole Slightly off-white, crystalline powder MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Imidazole Slightly off-white, crystalline powder MP Biomedicals
Have Questions?
Catalog Number
Quantity
MPB0210203301
1 kg
Catalog number
MPB0210203301
Price (SGD)
361.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (SGD)
361.00
EA
Add to cart
Specifications
Chemical Name or Material
Imidazole
CAS
288-32-4
Color
White
Density
500 - 600 kg/m3 (bulk density)
Molecular Formula
C3H4N2
View more