Check Order Status
Diastase
MP Biomedicals
Diastase
Malt
Have Questions?
Change view
Catalog Number
Quantity
MPB0210153980
100 g
MPB0210153925
25 g
2 Options
Catalog number
MPB0210153980
Price (SGD)
255.00
EA
Add to cart
Quantity:
100 g
Price (SGD)
255.00
EA
Add to cart
Diastase, [9000-92-4]
Activity: 1000 Lintner u/g
Melting Point: 175°C (dec)
Specifications
Concentration
1000 U/g
Color
Off-White
For Use With (Application)
Digests 50g of Starch in <30 min.
Source
Malt
Quantity
100 g
Product Type
Diastase
Form
Powder
Unit Size
EA
Contents & Storage
4°C