Check Order Status
Dextran Sulfate Sodium Salt (6,000-8,000 M.Wt), MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Dextran Sulfate Sodium Salt (6,000-8,000 M.Wt), MP Biomedicals
Have Questions?
Catalog Number
Quantity
MPB0210151890
500 g
Catalog number
MPB0210151890
Price (SGD)
7,657.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (SGD)
7,657.00
EA
Add to cart
Specifications
Chemical Name or Material
DEXTRAN SULFATE SODIUM SALT (6,000-8,000 M.Wt)
Color
White
CAS
9011-18-1
Physical Form
Powder
Recommended Storage
Store at Room Temperature (15-30°C)
View more
Dextran Sulfate Sodium (DSS) which has proven to have versatile value in Biomedical/clinical research
DSS can be used as an anticoagulant, antiviral as well as antilipemic agent