Dextran Sulfate (6000 to 8000), MP Biomedicals™
Dextran Sulfate (6000 to 8000), MP Biomedicals™
Dextran Sulfate (6000 to 8000), MP Biomedicals™
Dextran Sulfate (6000 to 8000), MP Biomedicals™
MP Biomedicals
Dextran Sulfate (6000 to 8000), MP Biomedicals™
Used as an anticoagulant, antiviral and antilipemic agent
Have Questions?
Catalog number MPB0210151850
Price (SGD)
1,056.00
Each
Add to cart
Request bulk or custom format
Price (SGD)
1,056.00
Each
Add to cart
Chemical Identifiers
CAS9011-18-1
Specifications
Chemical Name or MaterialDextran sulfate
ColorWhite to Off-white
pH5.0 to 7.5 (1% aqueous solution)
CAS9011-18-1
Physical FormPowder
View more
Dextran sulfate is a polyanionic derivative of dextran produced by esterification of Dextran with chlorosulphonic acid. Dextran sulfate sodium (DSS) is the sodium salt which is a white to off-white powder freely soluble in water and salt solutions to form a stable, clear solution.
  • Sulfur content is approximately 17% which corresponds to an average of 1.9 sulfate groups per glucosyl residue of the dextran molecule
  • High purity and consistent quality enables its application in pharmaceuticals and biotechnology industry
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.