Dextran Sulfate Sodium Salt Reagent MP Biomedicals
Dextran Sulfate Sodium Salt Reagent MP Biomedicals
Dextran Sulfate Sodium Salt Reagent MP Biomedicals
Dextran Sulfate Sodium Salt Reagent MP Biomedicals
MP Biomedicals
Dextran Sulfate Sodium Salt Reagent MP Biomedicals
Have Questions?
Catalog NumberQuantity
MPB02101516011 g
Catalog number MPB0210151601
Price (SGD)
138.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (SGD)
138.00
EA
Add to cart
Specifications
Chemical Name or MaterialDextran Sulfate Sodium Salt
CAS9011-18-1
Molecular Formula(C6H7O5)m(C6H7O5)n
MDL NumberMFCD00081551
Synonymbenzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
View more