Check Order Status
Choline chloride, 99%, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Choline chloride, 99%, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0210138601
1 kg
Catalog number
MPB0210138601
Price (SGD)
205.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (SGD)
205.00
EA
Add to cart
Specifications
Chemical Name or Material
Choline chloride
CAS
67-48-1
Color
White
Density
1.1g/cm
3
at 20°C
Assay Percent Range
0.99
View more