Cholesterol, NF Grade, MP Biomedicals
Cholesterol, NF Grade, MP Biomedicals
Cholesterol, NF Grade, MP Biomedicals
Cholesterol, NF Grade, MP Biomedicals
MP Biomedicals
Cholesterol, NF Grade, MP Biomedicals
Have Questions?
Catalog NumberQuantity
MPB02101380.325 g
Catalog number MPB02101380.3
Price (SGD)
76.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (SGD)
76.00
EA
Add to cart
Specifications
Chemical Name or MaterialCHOLESTEROL
CAS57-88-5
Melting Point147°C to 150°C
ColorWhite
Recommended Storage-20°C.
View more
  • Cholesterol is suitable for use in liposome preparations
  • Cholesterol is a component used for the preparation and study of artificial model membranes
  • One example is a study of valinomycin induced changes in membrane potentials of red blood cell and phospholipid (phosphatidylcholine from egg yolk plus cholesterol) vesicle suspensions
  • Positively charged cyanine dyes were used that fluorimetrically respond to the change in potential