Check Order Status
D(+)-Cellobiose; ≥99%, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
D(+)-Cellobiose; ≥99%, MP Biomedicals™
Cellobiose is a disaccharide composed of two glucose molecules; These are β-1,4-glycosidically linked. It is reducing sugar, which is readily soluble in polar solvents such as water and ethanol
Have Questions?
Catalog Number
Quantity
MPB0210129880
100 g
Catalog number
MPB0210129880
Price (SGD)
496.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (SGD)
496.00
EA
Add to cart
Specifications
Chemical Name or Material
D(+)-Cellobiose
CAS
528-50-7
Color
White
Recommended Storage
Store at Room Temperature
EINECS Number
208-436-5
View more