Check Order Status
β-Nicotinamide adenine dinucleotide, reduced, disodium salt, >99%, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
β-Nicotinamide adenine dinucleotide, reduced, disodium salt, >99%, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0210116801
1 g
Catalog number
MPB0210116801
Price (SGD)
246.00
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (SGD)
246.00
Each
Add to cart
Specifications
Chemical Name or Material
β-Nicotinamide adenine dinucleotide, reduced, disodium salt
CAS
606-68-8
Color
White to Yellow
Assay Percent Range
>99%
Molecular Formula
C21H27N7Na2O14P2
View more