Ribonuclease A, From Bovine Pancreas
Product Image
MP Biomedicals
Ribonuclease A, From Bovine Pancreas
Aggregates can be converted to monomers.
Have Questions?
Catalog NumberQuantity
MPB0210107680100 mg
Catalog number MPB0210107680
Price (SGD)
181.00
Each
Add to cart
Quantity:
100 mg
Price (SGD)
181.00
Each
Add to cart
Ribonuclease A, [9001-99-4]
  • RNase A
  • From Bovine Pancreas
  • Prepared from aggregate-free RNase
  • Lyophilized
  • Activity: 50 Kunitz units/mg.
Specifications
Concentration≥50 kU/mg
ColorWhite
Purity>70%
pH7 to 7.5
For Use With (Application)Used to remove RNA from DNA plasmid preparations and Protein samples
SourceBovine pancreas
Quantity100 mg
Product TypeRibonuclease A
Molecular Weight (g/mol)13700
InhibitorsHeavy Metal ions and is competitively inhibited by DNA
FormLyophilized Powder
Unit SizeEach
Contents & Storage
0°C