D-Biotin, MP Biomedicals
D-Biotin, MP Biomedicals
D-Biotin, MP Biomedicals
D-Biotin, MP Biomedicals
MP Biomedicals
D-Biotin, MP Biomedicals
Have Questions?
Catalog NumberQuantity
MPB02101023.1100 mg
Catalog number MPB02101023.1
Price (SGD)
72.00
EA
Add to cart
Quantity:
100 mg
Request bulk or custom format
Price (SGD)
72.00
EA
Add to cart
Specifications
Chemical Name or MaterialD-BIOTIN
CAS58-85-5
Melting Point223°C to 233°C
ColorWhite
Recommended StorageStore at +4°C.
View more
  • D-Biotin may be used to elute proteins from avidin/streptavidin resins
  • It is widely used for dietary supplements and fortified foods
  • It is also used for tablets and hard-shell capsule preparation due to its pharmaceutical properties