Check Order Status
L-Ascorbic acid, ≥98%, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
L-Ascorbic acid, ≥98%, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0210076925
25 g
Catalog number
MPB0210076925
Price (SGD)
82.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (SGD)
82.00
EA
Add to cart
Specifications
Chemical Name or Material
L-Ascorbic acid
CAS
50-81-7
Storage Note 1
Protect from light.
Color
White to Yellow
Recommended Storage
Store at room temperature.
View more