Hyaluronidase, Bovine Testes
Hyaluronidase, Bovine Testes
Actual product may vary
MP Biomedicals
Hyaluronidase, Bovine Testes
Bovine Testes
Have Questions?
Catalog NumberQuantity
MPB0210074080100 mg
Catalog number MPB0210074080
Price (SGD)
166.00
EA
Add to cart
Quantity:
100 mg
Price (SGD)
166.00
EA
Add to cart
Hyaluronidase, [37326-33-3]
  • Hyaluronate Glycanohydrolase
  • Activity: 300 USP units/mg min.
  • Lyophilized powder
Specifications
Concentration≥300 U/mg
ColorTan
pH4.5 to 6
For Use With (Application)Drug dispersion and delivery
SourceBovine testes
Quantity100 mg
Product TypeHyaluronidase
Molecular Weight (g/mol)≈55
InhibitorsFe2+ and Fe3+ are inhibitory as are Mn2+ and Cu2+
FormLyophilized Powder
Unit SizeEA
Contents & Storage
-20°C