Elastase 2X Crystallized, from Porcine Pancreas
Product Image
MP Biomedicals
Elastase 2X Crystallized, from Porcine Pancreas
Activity 50 u/mg protein, Porcine Pancreas, Milky Solution
Have Questions?
Catalog NumberQuantity
MPB021006171010 mg
Catalog number MPB0210061710
Price (SGD)
126.00
EA
Add to cart
Quantity:
10 mg
Price (SGD)
126.00
EA
Add to cart
Elastase was used to study human emphysema by induction of emphysema in rodents. It has been used in a study to investigate the design, synthesis and evaluation of biomimetic affinity ligands for elastases. Elastase from porcine pancreas has also been used in a study to investigate the purification and partial characterization of the pancreatic proteolytic enzymes trypsin, chymotrypsin, and elastase. It hydrolyzes peptide bonds, especially those adjacent to neutral amino acids.
Specifications
Concentration2X
ColorMilky
For Use With (Application)Investigate the purification and partial characterization of the pancreatic proteolytic Enzymes
SourcePorcine pancreas
Quantity10 mg
Product TypeElastase
Molecular Weight (g/mol)≈25000
InhibitorsDFP, Elastinal, alpha-2-macroglobulin
FormSolution
Unit SizeEA
Contents & Storage
4°C