▸
Organic compounds
▸
Unclassified Organic Compounds
▸
DL-Dithiothreitol, ≥99.0%, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
DL-Dithiothreitol, ≥99.0%, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0210059705
5 g
Catalog number
MPB0210059705
Price (SGD)
233.00
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (SGD)
233.00
Each
Add to cart
Specifications
Chemical Name or Material
DL-Dithiothreitol
CAS
27565-41-9
Color
White
EINECS Number
248-531-9
Assay Percent Range
≥99.0%
View more