Streptomycin Sulfate, White Powder, MP Biomedicals™
Streptomycin Sulfate, White Powder, MP Biomedicals™
Streptomycin Sulfate, White Powder, MP Biomedicals™
Streptomycin Sulfate, White Powder, MP Biomedicals™
MP Biomedicals
Streptomycin Sulfate, White Powder, MP Biomedicals™
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
25 g
Catalog number MPB0210055650
Price (SGD)
85.00
EA
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (SGD)
85.00
EA
Add to cart
Specifications
Chemical Name or MaterialStreptomycin Sulfate
TypeStreptomycin Sulfate
Physical FormSolid
CAS3810-74-0
Recommended StorageStore at 4°C.
View more