Mitomycin C, MP Biomedicals™
Mitomycin C, MP Biomedicals™
Mitomycin C, MP Biomedicals™
Mitomycin C, MP Biomedicals™
MP Biomedicals
Mitomycin C, MP Biomedicals™
The most widely used Mitomycin antibiotic. Mitomycin C, MP Biomedicals is the least toxic of the mitomycin antibiotics, with the most potent antitumor activity. It is an inhibitor of DNA synthesis, nuclear division, and cancer cells.
Have Questions?
Change viewbuttonViewtableView
Quantity:
20 mg
2 mg
10 mg
Catalog number MPB0210049820
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
20 mg
Request bulk or custom format
Specifications
CAS50-07-7
pH6 to 9 (Stability)
Extinction CoefficientEmM(367 nm) = 21.84 (water); EmM(357 nm) = 22.78 (methanol); E1% (216 nm) = 742 (methanol); E1% (360 nm) = 742 (methanol); E1% (560 nm) = 0.06 (methanol) (Lit.)
Recommended StorageStore at +4°C.
Potency850μg/mg minimum
View more
  • Antibacterial to gram positive, gram negative and acid-fast bacilli
  • Decomposes DNA in cancer cells by forming a complex with DNA
  • Inhibits division of cancer cells by interfering with the biosynthesis of DNA