Check Order Status
Acetyl Coenzyme A Trilithium salt Trihydrate, >92%, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Acetyl Coenzyme A Trilithium salt Trihydrate, >92%, MP Biomedicals™
Have Questions?
Change view
Quantity:
25 mg
1 mg
Catalog number
MPB0210049025
Price (SGD)
552.00
EA
Add to cart
Quantity:
25 mg
Request bulk or custom format
Price (SGD)
552.00
EA
Add to cart
Specifications
Chemical Name or Material
Acetyl Coenzyme A Trilithium Salt Trihydrate
CAS
75520-41-1
Color
White
Recommended Storage
Store at -20°C.
Assay Percent Range
>92%
View more