α-amylase Powder
Actual product may vary
MP Biomedicals
α-amylase Powder
α-amylase powder from Bacillus subtilis for the enzymatic liquefaction of starch.
Have Questions?
Catalog NumberQuantity
MPB0210044780100 g
Catalog number MPB0210044780
Price (SGD)
204.00
Each
Quantity:
100 g
Price (SGD)
204.00
Each
α-amylase powder from Bacillus subtilis for the enzymatic liquefaction of starch.
α-amylase powder from Bacillus subtilis is a liquefying diastase supplied as a dry, free-flowing powder with high uniformity and storage stability. One unit will dextrinize 1 mg of starch per minute at pH 6.6 and 30°C.
  • Activity: 500 to 1000 units/mg protein
  • ∼165,000 BAU/g
  • Deactivation temperature: 90°C to 100°C
  • For enzymatic liquefaction of starch
Specifications
CAS9000-90-2
Donor SourceBacillus subtilis
FormPowder
For Use With (Application)Carbohydrate Hydrolysis
Enzymeα-amylase
Quantity100 g
Unit SizeEach