Check Order Status
Catalase
Actual product may vary
MP Biomedicals
Catalase
Fungal suspension from
Aspergillus niger
Have Questions?
Catalog Number
Quantity
MPB0210040280
100 mL
Catalog number
MPB0210040280
Price (SGD)
283.00
EA
Add to cart
Quantity:
100 mL
Price (SGD)
283.00
EA
Add to cart
Catalase, [9001-05-2]
Density: 1.2g/mL
Activity: 1000 units/ml
Specifications
Concentration
≥1000 U/mL
For Use With (Application)
Removal of Peroxides, the generation of Oxygen and in coupled systems, for the determination of metabolites
Source
Aspergillus niger
Quantity
100 mL
Product Type
Catalase
Molecular Weight (g/mol)
≈250
Form
Liquid
Unit Size
EA
Contents & Storage
4°C