Check Order Status
Cycloheximide from Microbial, MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Cycloheximide from Microbial, MP Biomedicals
Have Questions?
Catalog Number
Quantity
MPB0210018325
25 g
Catalog number
MPB0210018325
Price (SGD)
1,454.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (SGD)
1,454.00
EA
Add to cart
Specifications
Chemical Name or Material
CYCLOHEXIMIDE from Microbial
CAS
66-81-9
Melting Point
103°C to 115°C
Color
White
Recommended Storage
+4°C
View more
Cycloheximide is used in protein synthesis in apoptosis
It is also used in plant research to study disease resistance and as an ethylene stimulant, useful in studies involving fruit and leaf production