Check Order Status
Acetylcholine chloride, ∽99%, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Acetylcholine chloride, ∽99%, MP Biomedicals™
Acetylcholine chloride is a cholinergic neurotransmitter that can induce the opening of calcium channels, as well as act on nicotinic and muscarinic acetylcholine receptors
Have Questions?
Catalog Number
Quantity
MPB0210001225
25 g
Catalog number
MPB0210001225
Price (SGD)
210.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (SGD)
210.00
EA
Add to cart
Specifications
Chemical Name or Material
Acetylcholine chloride
CAS
60-31-1
Color
White
EINECS Number
200-468-8
Assay Percent Range
∽99%
View more