▸
Solvents
▸
Other Solvents
▸
Liquid Scintillation Counting Cocktails and Solvents
▸
Ecolite(+) LC Fluid, MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Ecolite(+) LC Fluid, MP Biomedicals
Liquid Scintillation Cocktails; Ecolite(+); MP Biomedical; Biodegradable
Have Questions?
Catalog Number
Quantity
MPB0188247504
4 x 1 gal.
Catalog number
MPB0188247504
Price (SGD)
1,747.00
Each
Add to cart
Quantity:
4 x 1 gal.
Request bulk or custom format
Price (SGD)
1,747.00
Each
Add to cart
Specifications
Chemical Name or Material
Liquid Scintillation Cocktails
Quantity
4 x 1 gal.