Check Order Status
Water, Ultra Trace Elemental Analysis Grade, Fisher Chemical™
Element Search
Structure Search
Water, Ultra Trace Elemental Analysis Grade, Fisher Chemical™
Packaged in specially precleaned LDPE bottles to protect product integrity
Have Questions?
Change view
Quantity:
2 L
1 L
Packaging:
LDPE Bottle
Catalog number
MICW-92
Price (SGD)
750.00
EA
Add to cart
Quantity:
2 L
Packaging:
LDPE Bottle
Request bulk or custom format
Price (SGD)
750.00
EA
Add to cart
Specifications
Chemical Name or Material
Water
pH
7
CAS
7732-18-5
ChemAlert Storage Symbol
Gray
Color
Colorless
View more
Evaluated for 65 metal impurities in ppt range
Also specified for ≤1ppb for chloride, phosphate and sulfate anion impurities
Packaged in precleaned LDPE bottles to maintain product integrity