Grip Seal Bags
Grip Seal Bags
Actual product may vary
Grip Seal Bags
Provides storage for large number of samples
Have Questions?
Catalog NumberDimensions (L x W)
MICW-23 x 7.5 in.
Catalog number MICW-2
Price (SGD)
-
Dimensions (L x W):
3 x 7.5 in.
  • Cost effective sampling
  • Sample storage of large numbers of samples
Specifications
AutoclavableNot Autoclavable
Closure TypeGrip Seal
ColorTransparent
DisposableSingle-use
For Use With (Application)Sampling
MaterialPolyethylene
SterilityNon-sterile
Thickness (Metric)50 μm
Film DescriptionPolyethylene
No. per Case1000
No. per Pack1000
Outer PackagingBox
Quantity1000
Dimensions (L x W)3 x 7.5 in.
Product TypeSample Bag
Unit SizeEA
Contents & Storage
Room Temperature