▸
Facility Safety and Maintenance
▸
Cleaning Supplies and Equipment
▸
Mops and Mop Accessories
▸
Controlled Environments Mops, Mop Heads, and Handles
▸
TwistMop Cleanroom Mop
Micronova
TwistMop Cleanroom Mop
Handle grips turn in opposite directions to wring mop heads
Have Questions?
Catalog Number
Material
MICTM-9
Polyester
Catalog number
MICTM-9
Price (SGD)
-
Material:
Polyester
TwistMop Handle
Aluminum handle
Delrin and polypropylene fittings
Autoclavable
Attaches to either Polyester (TM-9) or PVA (TM-1) TwistMop Heads
Polyvinyl Alcohol (PVA) TwistMop Heads (TM-1)
Self-wringing mop head
Fast-drying and absorbent
Ideal for cleanroom support and cleanroom construction
Not compatible with Isoproppyl Alcohol (IPA)
Polyester TwistMop Heads (TM-9)
Self-wringing looped polyester mop head
Ideal for laying down disinfectant
Launderable
Autoclavable
Specifications
Product Type
Twist Mop Head
For Use With (Equipment)
Twist Mop Cleanroom Mop
For Use With (Application)
Ideal for Cleanroom Support and Cleanroom
Material
Polyester